
Triphenyl phosphate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:115-86-6
:
Formula:C₁₈H₁₅O₄P
MDL Number:MFCD00003031
:326.29 g/mol
Merck Index:12,09872
Boiling Pt:412 °C (1013 hPa)
Melting Pt:50…51 °C
Density:1.33 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-112-2
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
Flash Pt:223 °C
:GHS09
Specification Test Results
- Solubility:0.0019 g/l (25 °C)
- Melting Point:48 - 50 °C
- Molar Mass:326.28 g/mol
- Bulk Density:650 kg/m3
- Boiling Point:220 °C (5 hPa)
- Vapor Pressure:- 0.01 hPa (20 °C)
- Flash Point:230 °C
- Ignition Point:500 °C
Specification Test Results
Frequently Bought Together