Diphosphorus pentoxide ≥99.99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1314-56-3
:
Formula:P₂O₅
MDL Number:MFCD00011440
:141.94 g/mol
Merck Index:14,7355
Boiling Pt:360 °C (subl.)
Melting Pt:340 °C
Density:2.39
Storage Temperature:Ambient
EINECS:215-236-1
UN:1807
ADR:8,II
:GHS05
Related Information
Frequently Bought Together