Order Entry
Portugal
Orders LinkContactUsLinkComponent
Silicon dioxide, Sigma-Aldrich®
 :   Merck
Silicon dioxide, Sigma-Aldrich®
 :  Merck
Silicon dioxide, Sigma-Aldrich®
 :  Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
  •  :7631-86-9
  •  :SiO₂
  •  :MFCD00011232
  •  :60.08 g/mol
  •  :14,08493
  •  :2230 °C (1013 hPa)
  •  :1719 °C
  •  :2.2 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  •  :Ambient
  •  :231-545-4
Product Family Options
  • Pack type
    Pk
  • Glass bottle for solids
    50 g
    1 * 50 g
    1 * 50 g - 254,00 €
    254,00 €
    1 * 50 g
       254,00 €
  • Glass bottle for solids
    250 g
    1 * 250 g
    1 * 250 g - 918,00 €
    918,00 €
    1 * 250 g
       918,00 €
  • Glass bottle for solids
    500 g
    1 * 500 g
    1 * 500 g - 492,00 €
    492,00 €
    1 * 500 g
       492,00 €