
Germanium(IV) oxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1310-53-8
:
Formula:GeO₂
MDL Number:MFCD00011030
:104.61 g/mol
Merck Index:12,04417
Melting Pt:1110 °C
Density:3.64 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:215-180-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together