Order Entry
Portugal
ContactUsLinkComponent
Aluminium oxide, α-phase, Sigma-Aldrich®
Aluminium oxide, α-phase, Sigma-Aldrich®
Catalog # SIALBCR169-60G
Supplier:  Merck
CAS Number:  
Aluminium oxide, α-phase, Sigma-Aldrich®
Catalog # SIALBCR169-60G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    60 g
  • Seq:
    3377

Specifications

About this item

 :
 :101.96 g/mol
EINECS:215-691-6
CAS Number:1344-28-1
Formula:Al₂O₃
MDL Number:MFCD00003424