Order Entry
Portugal
ContactUsLinkComponent
 
Calcium hydroxide ≥98.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 10304KA
Supplier:  VWR Chemicals
CAS Number:  
undefined
Calcium hydroxide ≥98.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 10304KA
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Cardboard carton
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay:
    Min. 98.0 %
  • Insoluble in hydrochloric acid:
    Max. 0.02 %
  • Cl (Chloride):
    Max. 0.1 %
  • SO4 (Sulphate):
    Max. 0.01 %
  • Total N (Nitrogen):
    Max. 0.005 %
  • As (Arsenic):
    Max. 0.0001 %
  • Ba (Barium):
    Max. 0.005 %
  • Cu (Copper):
    0.001 %
  • Fe (Iron):
    Max. 0.005 %
  • K (Potassium):
    Max. 0.05 %
  • Mg (Magnesium):
    Max. 0.05 %
  • Na (Sodium):
    Max. 0.05 %
  • Pb (Lead):
    Max. 0.001 %
  • Sr (Strontium):
    Max. 0.05 %

Specification Test Results

About this item

 :
 :74.09 g/mol
Merck Index:13,01675
Melting Pt:550 °C
Density:2.24 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-137-3
UN:3262
ADR:8,III
 :GHS05
CAS Number:1305-62-0
Formula:Ca(OH)₂
MDL Number:MFCD00010901