Order Entry
Portugal
ContactUsLinkComponent
Calcium oxide ≥92%, powder, TECHNICAL
Calcium oxide ≥92%, powder, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Calcium oxide ≥92%, powder, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 92 %

Specification Test Results

Frequently Bought Together