Calcium hydride, Sigma-Aldrich®
Catalog # SIAL558257-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:42.09 g/mol
Merck Index:13,01674
Melting Pt:816 °C
Density:1.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-189-2
UN:1404
ADR:4.3,I
:M-Clarity™ quality level = MQ100
:GHS02
CAS Number:7789-78-8
Formula:H₂Ca
MDL Number:MFCD00010897