Calcium hydride ≥92%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 92% min CAS No.: 7789-78-8 Molecular Formula: CaH2 Formula Weight: 42.10 Physical Form: 1-20mm Granules Density: 1.900 Melting Point: 600° dec. Solubility: Decomposes in water, alcohol and acids to H2 Merck Reference: 14,1672 UN No.: 1404 MDL No.: MFCD00010897
Synonyms: Calcium dihydride
- CAS Number:7789-78-8
- Formula:H₂Ca
- MDL Number:MFCD00010897
- Molecular Weight:42.09 g/mol
- Merck Index:14,1672
- Melting Pt:600 °C (dec.)
- Density:1.900
- Storage Temperature:Ambient
- EINECS:232-189-2
- UN:1404
- ADR:4.3,I
2 Options Available Below
Product Details & Documents
Purity: 92% min CAS No.: 7789-78-8 Molecular Formula: CaH2 Formula Weight: 42.10 Physical Form: 1-20mm Granules Density: 1.900 Melting Point: 600° dec. Solubility: Decomposes in water, alcohol and acids to H2 Merck Reference: 14,1672 UN No.: 1404 MDL No.: MFCD00010897
Synonyms: Calcium dihydride
- CAS Number:7789-78-8
- Formula:H₂Ca
- MDL Number:MFCD00010897
- Molecular Weight:42.09 g/mol
- Merck Index:14,1672
- Melting Pt:600 °C (dec.)
- Density:1.900
- Storage Temperature:Ambient
- EINECS:232-189-2
- UN:1404
- ADR:4.3,I



