Calcium hydride ∼ 93%, grains 0-20 mm, extra pure (up to 15% powder)
Catalog # ACRO196791000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Appearance:Grey Granules
- Assay:≥92 % (Total Ca)
- Particle size:1 to 20 mm
- Aluminium (Al):≤100 ppm
- Hydrogen (H):980 ml/g CaH2
- Iron (Fe):≤100 ppm
- Magnesium (Mg):≤8000 ppm
- Nitrogen (N):≤2000 ppm
Specification Test Results
About this item
:
:42.09 g/mol
Merck Index:13,01674
Melting Pt:675 °C
Density:1,9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-189-2
UN:1404
ADR:4.3,I
:GHS02
:GHS07
CAS Number:7789-78-8
Formula:H₂Ca
MDL Number:MFCD00010897