Calcium fluoride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7789-75-5
:
Formula:CaF₂
MDL Number:MFCD00010907
:78.07 g/mol
Merck Index:13,01669
Boiling Pt:2500 °C (1013 hPa)
Melting Pt:1418 °C
Density:3.18 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:232-188-7
:GHS07
Frequently Bought Together