Barium chromate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10294-40-3
:
Formula:BaCrO₄
MDL Number:MFCD00014180
:253.32 g/mol
Merck Index:12,00999
Melting Pt:1400 °C
Density:4.5 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:233-660-5
ADR:5.1,III
:M-Clarity™ quality level = MQ200
UN:1479
:GHS03
:GHS07
:GHS09
Frequently Bought Together