Barium chromate ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10294-40-3
:
Formula:BaCrO₄
MDL Number:MFCD00014180
:253.32 g/mol
Merck Index:14,972
Melting Pt:1400 °C
Density:4.50
EINECS:233-660-5
UN:1479
ADR:5.1,III
:GHS08
:GHS03
:GHS07
Storage Temperature:Ambient
Frequently Bought Together