Order Entry
Portugal
ContactUsLinkComponent
Calcium carbonate, Supelco®
Calcium carbonate, Supelco®
Catalog # SUPL21067-25G
Supplier:  Merck
CAS Number:  
Calcium carbonate, Supelco®
Catalog # SUPL21067-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :100.09 g/mol
Merck Index:13,01658
Melting Pt:825 °C
Density:2,71 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-439-9
CAS Number:471-34-1
Formula:CaCO₃
MDL Number:MFCD00010906