Order Entry
Portugal
ContactUsLinkComponent
Calcium carbonate ≥95%, powder, TECHNICAL
Calcium carbonate ≥95%, powder, TECHNICAL
Catalog # 22290.363
Supplier:  VWR Chemicals
CAS Number:  
undefined
Calcium carbonate ≥95%, powder, TECHNICAL
Catalog # 22290.363
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Assay:
    Min. 95 %

Specification Test Results

About this item

 :
 :100.09 g/mol
Merck Index:13,01658
Melting Pt:825 °C
Density:2.71 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-439-9
REACH:01-2119486795-18
CAS Number:471-34-1
Formula:CaCO₃
MDL Number:MFCD00010906