Calcium carbonate, Certipur®, Supelco®
Catalog # 1.02410.0050
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Solubility:0.014 g/l (20 °C)
- Melting Point:825 °C (decomposition)
- Molar Mass:100.09 g/mol
- Bulk Density:400 - 1400 kg/m3
- pH:9.5 - 10.5 (100 g/l, H2O, 20 °C) (slurry)
Specification Test Results
About this item
:
:100.09 g/mol
Merck Index:13,01658
Melting Pt:825 °C
Density:2.71 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:207-439-9
CAS Number:471-34-1
Formula:CaCO₃
MDL Number:MFCD00010906