Order Entry
Portugal
ContactUsLinkComponent
Sodium orthosilicate (Mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution)
Corrosive
Irritant
Sodium orthosilicate (Mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution)
Catalog # 17568.A1
CAS Number:  
Corrosive
Irritant
Sodium orthosilicate (Mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution)
Catalog # 17568.A1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

CAS Number:13472-30-5
 :
Formula:Na₄O₄Si
MDL Number:MFCD00014252
 :184.04 g/mol
Melting Pt:1078 °C
Density:2.5 g/cm³ (26 °C)
Storage Temperature:Ambient
EINECS:236-741-3
UN:1759
ADR:8,II
 :GHS05
 :GHS07