
Sulphuric acid 95.0-98.0% ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Merck Index:14,08974
Boiling Pt:310 °C (1.013 hPa)
Melting Pt:–14…–10 °C
Density:1.840 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
:GHS05
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Vapor Density:<0.3 (25 °C, vs air)
- Assay:95.0 - 98.0 %
- Density (D 20/4):1.825 - 1.836
- Residue on ignition:Max. 5 ppm
- Ammonium (NH₄):Max. 2 ppm
- Arsenic (As):Max. 0.01 ppm
- Iron (Fe):Max. 0.2 ppm
- Mercury (Hg):Max. 0.005 ppm
- Heavy metals (as Pb):Max. 1 ppm
- Chloride (Cl):Max. 0.2 ppm
- Nitrate (NO₃):Max. 0.5 ppm
- KMnO₄ reducing matter (as SO₂):Max. 2 ppm
- APHA:Max. 10
- Appearance:complying
- Vapor Pressure:1 mmHg ( 146 °C)
Specification Test Results
Frequently Bought Together