Order Entry
Portugal
Orders LinkContactUsLinkComponent
Cyclohexanone, Sigma-Aldrich®

Cyclohexanone, Sigma-Aldrich®

Flammable
Irritant
Supplier:   Merck
Cyclohexanone, Sigma-Aldrich®
Supplier:  Merck
Certificates
Cyclohexanone, Sigma-Aldrich®
Cyclohexanone, Sigma-Aldrich®

4 Options Available Below

Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • UN:1915
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ300
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • REACH:01-2119453616-35
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • 1
    Log in for availability
    Log in for availability
    1 * 1 unid.
    1 * 1 unid. - 64,00 €
    64,00 €
    1 * 1 unid.
       64,00 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 89,80 €
    89,80 €
    1 * 1 kg
       89,80 €

  • 10 kg
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 218,00 €
    218,00 €
    1 * 10 kg
       218,00 €

  • 25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - 207,00 €
    207,00 €
    1 * 25 kg
       207,00 €