Ethyl 4-hydroxybenzoate 99%, white powder, ReagentPlus®, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:120-47-8
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00002353
Merck Index:13,03869
Boiling Pt:297…298 °C (lit.)
Melting Pt:114…117 °C (lit.)
Density:1.078 g/cm³ (131 °C)
:M-Clarity™ quality level = MQ200
:166.18 g/mol
Storage Temperature:Ambient
EINECS:204-399-4
Frequently Bought Together