Order Entry
Portugal
Orders LinkContactUsLinkComponent
Vanillin, Sigma-Aldrich®

Vanillin, Sigma-Aldrich®

Irritant
Supplier:   Merck
Vanillin, Sigma-Aldrich®
Supplier:  Merck
Certificates
Vanillin, Sigma-Aldrich®
Vanillin, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,09998
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:∼82 °C
  • Density:1.056 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ300
  • Flash Pt:153 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 239,00 €
    239,00 €
    1 * 1 kg
       239,00 €

  • 10 kg
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 1 240,00 €
    1 240,00 €
    1 * 10 kg
       1 240,00 €

  • 25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - 2 360,00 €
    2 360,00 €
    1 * 25 kg
       2 360,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.