Vanillin ≥99%, off-white powder
Supplier: MP Biomedicals
Jump to:
About this item
Soluble in alcohol(50 mg/ml (5%); Ethanol), chloroform and ether; slightly soluble in water and glycol.
Soluble in alcohol(50 mg/ml (5%); Ethanol), chloroform and ether; slightly soluble in water and glycol.
Synonyms: 4-Hydroxy-3-methoxybenzaldehyde
- CAS Number:121-33-5
- Formula:C₈H₈O₃
- MDL Number:MFCD00006942
- Molecular Weight:152.15 g/mol
- Merck Index:13,09998
- Boiling Pt:285 °C (1013 hPa)
- Melting Pt:78…85 °C
- Density:1.056 g/cm³ (20 °C)
- Flash Pt:153 °C
- Storage Temperature:Ambient
- EINECS:204-465-2
3 Options Available Below
Product Details & Documents
Soluble in alcohol(50 mg/ml (5%); Ethanol), chloroform and ether; slightly soluble in water and glycol.
Soluble in alcohol(50 mg/ml (5%); Ethanol), chloroform and ether; slightly soluble in water and glycol.
Synonyms: 4-Hydroxy-3-methoxybenzaldehyde
- CAS Number:121-33-5
- Formula:C₈H₈O₃
- MDL Number:MFCD00006942
- Molecular Weight:152.15 g/mol
- Merck Index:13,09998
- Boiling Pt:285 °C (1013 hPa)
- Melting Pt:78…85 °C
- Density:1.056 g/cm³ (20 °C)
- Flash Pt:153 °C
- Storage Temperature:Ambient
- EINECS:204-465-2



