Order Entry
Portugal
ContactUsLinkComponent
trans-3-Chlorocinnamic acid for synthesis, Sigma-Aldrich®
Irritant
trans-3-Chlorocinnamic acid for synthesis, Sigma-Aldrich®
Catalog # 8.41324.0010
Supplier:  Merck
CAS Number:  
Irritant
trans-3-Chlorocinnamic acid for synthesis, Sigma-Aldrich®
Catalog # 8.41324.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Melting Point:
    163 - 165 °C
  • Molar Mass:
    182.6 g/mol

Specification Test Results

About this item

 :
 :182.6037 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:217-478-3
 :GHS07
CAS Number:14473-90-6
Formula:C₉H₇ClO₂
MDL Number:MFCD00004384