Isobutanol
Catalog #
Product Details & Documents
Synonyms: iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol
- CAS Number:78-83-1
- Formula:(CH₃)₂CHCH₂OH
- MDL Number:MFCD00004740
- Molecular Weight:74.12 g/mol
- Merck Index:13,05148
- Boiling Pt:107.9 °C (1013 hPa)
- Melting Pt:–108 °C
- Density:0.802 g/cm³ (20 °C)
- Flash Pt:28 °C
- Storage Temperature:Ambient
- EINECS:201-148-0
- UN:1212
- ADR:3,III



