

Dipropyl sulphate ≥90.0% (by GC) alkylating agent
: TCI
:598-05-0
:
:C₆H₁₄O₄S
:MFCD00059243
:182.24 g/mol
:115°C
:2922
:8,II
:GHS05
:GHS06
:Ambient
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 90.0 %
Frequently Bought Together