1,5-Diaminopentane 98%
:462-94-2
:
:C₅H₁₄N₂
:MFCD00008239
:102.18 g/mol
:12,01645
:178…180 °C (760 mmHg)
:9 °C
:0.87 g/cm³
:62 °C
:Ambient
:207-329-0
:2735
:8,II
:GHS05
:GHS07
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4570 to 1.4600 (20°C, 589 nm)
- GC:≥97.5 %
Frequently Bought Together