Order Entry
Portugal
ContactUsLinkComponent
Methyl trifluoromethanesulphonate for synthesis, Sigma-Aldrich®
Flammable
Corrosive
Methyl trifluoromethanesulphonate for synthesis, Sigma-Aldrich®
Catalog # 8.18035.0010
Supplier:  Merck
CAS Number:  
Flammable
Corrosive
Methyl trifluoromethanesulphonate for synthesis, Sigma-Aldrich®
Catalog # 8.18035.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Molar Mass:
    164.1 g/mol
  • Boiling Point:
    99 °C (1013 hPa)
  • Flash Point:
    38 °C
  • Refractive Index:
    1.3228 (25 °C, 589 nm)
  • Density:
    1.49 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :164.11 g/mol
Boiling Pt:95 °C (1013 hPa)
Density:1.45 g/cm³ (20 °C)
ADR:8 (3),II
 :M-Clarity™ quality level = MQ200
EINECS:206-371-7
UN:2920
 :GHS02
 :GHS05
Storage Temperature:Ambient
CAS Number:333-27-7
Formula:C₂H₃F₃O₃S
MDL Number:MFCD00000409