

Trifluoroacetic anhydride 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:407-25-0
:
Formula:C₄F₆O₃
MDL Number:MFCD00000416
:210.03 g/mol
Boiling Pt:39.1 °C (760 mmHg)
Melting Pt:–63.5 °C
Density:1.51 g/cm³
Storage Temperature:Ambient
EINECS:206-982-9
UN:3265
ADR:8,I
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- GC:≥99.0 %
- Chloride (Cl):≤50 ppm
- Fluoride (F):≤50 ppm
- Sulfate (SO4):≤500 ppm
Specification Test Results
Frequently Bought Together