

3-Mercaptopropionic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:107-96-0
:
Formula:C₃H₆O₂S
MDL Number:MFCD00004897
:106.15 g/mol
Boiling Pt:217 °C (1013 hPa)
Melting Pt:17…19 °C
Density:1.2199 g/cm³ (20 °C)
Flash Pt:93 °C
UN:2922
ADR:8 (6.1),II
:M-Clarity™ quality level = MQ200
EINECS:203-537-0
:GHS06
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:17 - 19 °C
- Molar Mass:106.14 g/mol
- Boiling Point:110 °C (20 hPa)
- Vapor Pressure:0.053 hPa (20 °C)
- Flash Point:93 °C
- Refractive Index:1.4911 (20 °C, 589 nm)
- Explosion Limit:1.60 %(V)
- Density:1.22 g/cm3 (20 °C)
- pH:2 (120 g/l, H2O, 20 °C)
- Ignition Point:350 °C
Specification Test Results
Frequently Bought Together