Order Entry
Portugal
Orders LinkContactUsLinkComponent
 

Urea

Supplier:   Promega
Urea
Supplier:  Promega
Certificates
Urea

2 Options Available Below

Product Details & Documents
Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:NH₂CONH₂
  • MDL Number:MFCD00008022
  • Molecular Weight:60.06 g/mol
  • Merck Index:13,09935
  • Boiling Pt:197 °C (1013 hPa)
  • Melting Pt:132.7 °C
  • Density:1.34 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-315-5
  • REACH:01-2119463277-33
Specifications for Product Family

Specification Test Results

  • Purity:
    ≥99.0%
  • Melting Point:
    132–135°C
  • A280 at 8M in water:
    ≤0.10
  • Chloride:
    ≤0.0005%
  • Heavy Metals:
    ≤0.001%
  • Iron:
    ≤0.001%
  • Cyanate:
    none detected
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 129,00 €
    129,00 €
    1 * 1 kg
       129,00 €

  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 408,00 €
    408,00 €
    1 * 5 kg
       408,00 €