Order Entry
Portugal
ContactUsLinkComponent
N,N-Dimethylacetamide 99.5+% for analysis
Irritant
Health
N,N-Dimethylacetamide 99.5+% for analysis
Catalog # ACRO390800010
CAS Number:  
undefined
Irritant
Health
N,N-Dimethylacetamide 99.5+% for analysis
Catalog # ACRO390800010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 %
  • GC:
    ≥99.50 %
  • Residue after evaporation:
    ≤0.0005 %
  • Acidity:
    ≤0.0005 meq/g
  • Chloride (Cl):
    ≤0.001 %
  • Sulfate (SO4):
    ≤0.001 %
  • Cadmium (Cd):
    ≤0.05 ppm
  • Calcium (Ca):
    ≤0.5 ppm
  • Chromium (Cr):
    ≤0.02 ppm
  • Copper (Cu):
    ≤0.02 ppm
  • Iron (Fe):
    ≤0.1 ppm
  • Lead (Pb):
    ≤0.1 ppm
  • Magnesium (Mg):
    ≤0.1 ppm
  • Nickel (Ni):
    ≤0.02 ppm
  • Zinc (Zn):
    ≤0.1 ppm

Specification Test Results

About this item

 :
 :87.12 g/mol
Merck Index:13,03254
Boiling Pt:164…166 °C (760 mmHg)
Melting Pt:–20 °C
Density:0.937 g/cm³
Flash Pt:70 °C
Storage Temperature:Ambient
EINECS:204-826-4
ADR:6.1,III
 :GHS07
 :GHS08
CAS Number:127-19-5
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686