(R)-(-)-2-Butanol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (R)-(-)-Ethyl methyl carbinol, (R)-(-)-sec-Butyl alcohol, (R)-(-)-2-Hydroxy butane, (R)-(-)-sec-Butanol, (R)-(-)-Methyl ethyl carbinol
CAS Number:14898-79-4
:
Formula:C₄H₁₀O
MDL Number:MFCD00064280
Boiling Pt:99…100 °C
Melting Pt:–114 °C
Density:0.806
Molecular Weight:74.12 g/mol
Flash Pt:26 °C (79 °F)
UN:1120
ADR:3 III
Merck Index:13,01540
EINECS:238-967-8
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together