(R)-(-)-2-Butanol 99%
: (R)-(-)-Ethyl methyl carbinol, (R)-(-)-sec-Butyl alcohol, (R)-(-)-2-Hydroxy butane, (R)-(-)-sec-Butanol, (R)-(-)-Methyl ethyl carbinol
:14898-79-4
:
:C₄H₁₀O
:MFCD00064280
:99…100 °C (760 mmHg)
:–114 °C
:0.8 g/cm³
:74.12 g/mol
:26 °C
:1120
:3 III
:13,01540
:238-967-8
:GHS07
:GHS02
:Ambient
- Environmentally Preferable:
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3960 to 1.3980 (20°C, 589 nm)
- GC:≥98.5 %
- Specific optical rotation:-12° to -14° (20°C, 589 nm) (neat)
Frequently Bought Together