Calcium sulphate dihydrate 99.0-102.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 22452.294
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:99.0 - 102.0 %
- NO3 (Nitrate):Passes test
- Heavy metals (as Pb):Max. 20 ppm
- Insolubility in hydrochloric acid:Max. 100 ppm
- Cl (Chloride):Max. 50 ppm
- Fe (Iron):Max. 10 ppm
Specification Test Results
About this item
:
:172.17 g/mol
Merck Index:13,01711
Density:2.32 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-900-3
CAS Number:10101-41-4
Formula:CaSO₄·2H₂O
MDL Number:MFCD00149625