Calcium sulphate hemihydrate ≥97.0%, Purum, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:10034-76-1
:26499-65-0
Formula:CaSO₄·0.5H₂O
MDL Number:MFCD00799067
Melting Pt:> 1400 °C
Density:2.320 g/cm³ (20 °C)
:145.15 g/mol
Storage Temperature:Ambient
EINECS:231-900-3
Specification Test Results
- Assay:Min. 97 %
Specification Test Results
Frequently Bought Together