Calcium bromide hydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:71626-99-8
:
Formula:Br₂Ca·H₂O
MDL Number:MFCD00149608
:217.9 g/mol
Density:2.295 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:232-164-6
Frequently Bought Together