Calcium bromide hydrate ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71626-99-8
:
Formula:Br₂Ca·H₂O
MDL Number:MFCD00149608
:217.9 g/mol
Density:2.295 g/cm³ (20 °C)
EINECS:232-164-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together