Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 1.000 mL
Catalog # PROMV42511000
Supplier: Promega
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Catalog # PROMV42511000
Supplier: Promega
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1.000 mL
- Description:TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



