Order Entry
Portugal
ContactUsLinkComponent
Hydrobromic acid 47%, AnalaR NORMAPUR® analytical reagent
Hydrobromic acid 47%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Hydrobromic acid 47%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    47.0 - 49.0 %
  • Heavy metals (as Pb):
    Max. 2 ppm
  • Ignition residue (SO4):
    Max. 50 ppm
  • Cl (Chloride):
    Max. 0.02 %
  • I (Iodide):
    Max. 30 ppm
  • PO4 + PO3 (as PO4):
    Max. 2 ppm
  • SO4 + SO3 (as SO4):
    Max. 30 ppm
  • As (Arsenic):
    Max. 0.5 ppm
  • Cd (Cadmium):
    Max. 0.5 ppm
  • Cu (Copper):
    Max. 0.5 ppm
  • Fe (Iron):
    Max. 1 ppm
  • Pb (Lead):
    Max. 0.5 ppm
  • Zn (Zinc):
    Max. 0.5 ppm

Specification Test Results

Frequently Bought Together