(±)-trans-1,2-Cyclohexanediol for synthesis, Sigma-Aldrich®
Catalog # 8.20366.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:375 g/l (20 °C)
- Melting Point:100 - 103 °C
- Molar Mass:116.15 g/mol
- Boiling Point:231 - 233 °C
- Vapor Pressure:4 hPa (110 °C)
- Flash Point:134 °C
- Ignition Point:360 °C
- Hill Formula:C6H12O2
- Chemical Formula:C6H10-1,2-(OH)2
Specification Test Results
About this item
:
Boiling Pt:237 °C (1013 hPa)
Melting Pt:100…103 °C
Density:1.147 g/cm³ (24 °C)
:M-Clarity™ quality level = MQ200
:116.16 g/mol
EINECS:215-956-6
CAS Number:1460-57-7
Formula:C₆H₁₂O₂
MDL Number:MFCD00063611