Tween® 20 (Polysorbate)
Supplier: Promega
Certificates
About this item
CAS Number:9005-64-5
:
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986
:1227.53 g/mol
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:500-018-3
Frequently Bought Together