Arsenic, pieces, Sigma-Aldrich®
Catalog # SIAL202657-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:74.92 g/mol
Merck Index:12,00832
Melting Pt:817 °C
Density:5.727 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-148-6
UN:1558
ADR:6.1,II
:M-Clarity™ quality level = MQ100
:GHS06
:GHS09
CAS Number:7440-38-2
Formula:As
MDL Number:MFCD00085309