Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Portugal
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
pro-Luciferin substrate
pro-Luciferin substrate 2J2/4F12 (Ester)
pro-Luciferin substrate 2J2/4F12 (Ester)
pro-Luciferin substrate 2J2/4F12 (Ester)
Catalog # PROMP1671
Supplier:
Promega
CAS Number:
Expand image
pro-Luciferin substrate 2J2/4F12 (Ester)
Catalog # PROMP1671
Supplier:
Promega
CAS Number:
Specifications
Pk:
3 mg
Seq:
2185
Specifications
About this item
Product Details
:
Product Details