0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Portugal
ContactUsLinkComponent
pro-Luciferin substrate
pro-Luciferin substrate 3A7
pro-Luciferin substrate 3A7
Catalog # PROMP1741
Supplier:
Promega
CAS Number:
Expand image
pro-Luciferin substrate 3A7
Catalog # PROMP1741
Supplier:
Promega
CAS Number:
Specifications
Pk:
3 mg
Seq:
2185
Specifications
About this item
Product Details
:
Product Details