Weighing funnels, PP
: Merck
These translucent weighing funnels are made of PP.
- Residual sample can be rinsed to guarantee 100% transfer
- Suitable for safe handling of hazardous materials
- Very good chemical and temperature resistance
Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer.
Frequently Bought Together