Human recombinant DLK1, HIS tag
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Notch ligand
Produced by HEK 293 cells. DLK1 (Ala24-Pro297) is fused at the C-terminus to a His-tag.
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Tag sequence:HIS Tag
- Protein synonyms:Pref-1|FA1|Protein delta homolog 1|Preadipocyte factor 1|Fetal antigen 1|pG2
- Protein/peptide name:DLK1
- Purity:>95% (SEC-HPLC and reducing SDS-PAGE)
- Formulation:Lyophilized. In 20 mM PBS, pH 7,2, containing 150 mM sodium chloride.
Specifications
Frequently Bought Together