Order Entry
Portugal
ContactUsLinkComponent
Human recombinant SUMO-3 (from E. coli), His-tag
Human recombinant SUMO-3 (from E. coli), His-tag
Catalog # ENZOBMLUW92150500
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant SUMO-3 (from E. coli), His-tag
Catalog # ENZOBMLUW92150500
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    500 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    SUMO-3
  • Purity:
    >95% (SDS-PAGE)
  • Formulation:
    Liquid. In 20 mM HEPES, pH 8,0, with 50 mM sodium chloride and 1 mM dithiothreitol.

Specifications

About this item

Produced in E. coli. Human SUMO-3 is fused to a His-tag.

Mature protein for functional studies.