Order Entry
Portugal
ContactUsLinkComponent
 
Human recombinant Otubain-1 (from E. coli), HIS Tag
Catalog # ENZOBMLUW06800100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant Otubain-1 (from E. coli), HIS Tag
Catalog # ENZOBMLUW06800100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    Otubain-1
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    1 mg/ml
  • Formulation:
    Liquid. In 50 mM TRIS, pH 7,5, containing 50 mM NaCl and 1 mM DTT.

Specifications

About this item

Produced in E. coli.