Mouse recombinant TRAIL (soluble) (from E. coli)
100 µG
Catálogo # ENZOBMLSE7220100
Fornecedor: Enzo Life Sciences
Mouse recombinant TRAIL (soluble) (from <i>E. coli</i>)
Catálogo # ENZOBMLSE7220100
Fornecedor: Enzo Life Sciences
Número CAS:
Ir para:
Product Details & Documents
Produced in E. coli. Soluble mouse TRAIL (aa 118-291).
Especificações
Especificações
- Pk:100 µG
- Cat. No.:ENZOBMLSE7220100
- Protein/peptide type:Recombinant
- Protein/peptide name:TRAIL (soluble)
- Species:Mouse
- Source:E. coli
- Biological activity:Induces apoptosis at 0,1 and 1,0 µg/ml, in primary cultured mouse hepatocytes, in the presence of cycloheximide (5 µg/ml) or actinomycin D (0,5 µg/ml), induces apoptosis in Jurkat cells (a TRAIL-sensitive human cell line) at 1 µg/ml, within 3 h, inhibits secondary clonal expansion of mouse CD8+ T cells in vitro (50 ng/ml and greater)
Especificações
Recomendações
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



