Human recombinant CDC25C (from E. coli)
Catalog # ENZOBMLSE3660050
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:CDC25C
Specifications
About this item
Produced in E. coli. Full length human CDC25C.